EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
2,4-Diaminopyrimidines as histamine H4 receptor ligands--Scaffold optimization and pharmacological characterization.
PMID 19773175
Abstract
The human histamine H(4) receptor (hH(4)R) is a promising new target in the therapy of inflammatory diseases and disorders of the immune system. For the development of new H(4)R antagonists a broad ligand-based virtual screening was performed resulting in two hits. The dissection of their common annelated aromatic core into its heteromonocyclic components showed that 2,4-diaminopyrimidine is a potent hH(4)R affinity scaffold, which was comprehensively investigated. Structure-activity relationship studies revealed that slight structural changes evoke extensive differences in functional activities and potencies: while o- and p-substituted benzyl amines mainly showed partial agonism, m-substituted and rigidified ones exhibited inverse agonist efficacy.