EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis, antiproliferative, and pharmacokinetic properties of 3- and 17-double-modified analogs of 2-methoxyestradiol.
PMID 19782564
Abstract
The syntheses of 21 analogs of 2-methoxyestradiol are presented, including ENMD-1198 which was selected for advancement into Phase 1 clinical trials in oncology. These analogs were evaluated for antiproliferative activity using breast tumor MDA-MB-231 cells, for antiangiogenic activity in HUVEC proliferation assays, and for estrogenic activity in MCF-7 cell proliferation. The most active analogs were evaluated for iv and oral pharmacokinetic properties via cassette dosing in rat and in mice pharmacokinetic models.