PloS one
Synergistic antibacterial and anti-inflammatory activity of temporin A and modified temporin B in vivo.
PMID 19784377
Abstract
Temporins are antimicrobial peptides secreted by the granular glands of the European red frog (Rana temporaria). They are 10-14 amino acid long polypeptides active prevalently against gram positive bacteria. This study shows that a synthetic temporin B analogue (TB-YK), acquires the capacity to act in synergism with temporin A and to exert antimicrobial and anti-inflammatory activity in vivo against gram positive and gram negative bacteria. Administration of 3.4 mg/Kg of temporin A (TA)+1.6 mg/Kg TB-YK, given to individual mice concurrently with a lethal dose of bacteria (gram positive or negative), rescued 100% of the animals. More importantly, the same doses of temporins, administered one week after experimental infection with a sub lethal dose of bacteria, sterilized 100% of the animals within 3-6 days. Also, it is described an animal model based on the use of sub lethal doses of bacteria, which closely mimics bacterial infection in humans. The model offers the possibility to test in a preclinical setting the true potential of TA and TB-YK in combination as antimicrobial and anti-inflammatory agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47636
Fmoc-Pro-OH, ≥99.0% (HPLC)
C20H19NO4
47638
Fmoc-Val-OH, ≥98.0% (HPLC)
C20H21NO4