EMAIL THIS PAGE TO A FRIEND
Proteome science
2-DE analysis indicates that Acinetobacter baumannii displays a robust and versatile metabolism.
PMID 19785748
Abstract
Acinetobacter baumannii is a nosocomial pathogen that has been associated with outbreak infections in hospitals. Despite increasing awareness about this bacterium, its proteome remains poorly characterised, however recently the complete genome of A. baumannii reference strain ATCC 17978 has been sequenced. Here, we have used 2-DE and MALDI-TOF/TOF approach to characterise the proteome of this strain. The membrane and cytoplasmatic protein extracts were analysed separately, these analyses revealed the reproducible presence of 239 and 511 membrane and cytoplamatic protein spots, respectively. MALDI-TOF/TOF characterisation identified a total of 192 protein spots (37 membrane and 155 cytoplasmatic) and revealed that the identified membrane proteins were mainly transport-related proteins, whereas the cytoplasmatic proteins were of diverse nature, although mainly related to metabolic processes. This work indicates that A. baumannii has a versatile and robust metabolism and also reveal a number of proteins that may play a key role in the mechanism of drug resistance and virulence. The data obtained complements earlier reports of A. baumannii proteome and provides new tools to increase our knowledge on the protein expression profile of this pathogen.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17978 Amberlite® CG-120-II, H+-form, strongly acidic, 200-400 mesh