EMAIL THIS PAGE TO A FRIEND
Journal of visualized experiments : JoVE
Preparation of rat brain aggregate cultures for neuron and glia development studies.
PMID 19794393
Abstract
An in vitro system that recapitulates the development and differentiation of progenitors into mature neurons and glia in the central nervous system (CNS) would provide a powerful platform for neuroscientists to investigate axo-glial interactions, properties and differentiation of multipotent progenitors, and progression of oligodendroglial lineage cells at the cellular and molecular level. We describe here a CNS aggregate culture system from embryonic rat forebrains, which can be maintained in a serum-free medium up to 3-4 weeks and is used in our laboratory as a model to study neuron-glia interaction and CNS myelination. This video clip will demonstrate how to isolate and grow these CNS aggregate cultures from E16 rat brain. Furthermore, from the same brain dissection, highly enriched regular dissociated neuronal cultures can be readily obtained and used for various studies on CNS neurons or used for co-cultures with other cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8783
Progesterone, powder, BioReagent, suitable for cell culture
C21H30O2