EMAIL THIS PAGE TO A FRIEND
Cancer cell
Proteomic and genetic approaches identify Syk as an AML target.
PMID 19800574
Abstract
Cell-based screening can facilitate the rapid identification of compounds inducing complex cellular phenotypes. Advancing a compound toward the clinic, however, generally requires the identification of precise mechanisms of action. We previously found that epidermal growth factor receptor (EGFR) inhibitors induce acute myeloid leukemia (AML) differentiation via a non-EGFR mechanism. In this report, we integrated proteomic and RNAi-based strategies to identify their off-target, anti-AML mechanism. These orthogonal approaches identified Syk as a target in AML. Genetic and pharmacological inactivation of Syk with a drug in clinical trial for other indications promoted differentiation of AML cells and attenuated leukemia growth in vivo. These results demonstrate the power of integrating diverse chemical, proteomic, and genomic screening approaches to identify therapeutic strategies for cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F2442
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
F6178
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture
F4135
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture
F0392
Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture
12107C
Fetal Bovine Serum, USA origin, γ-irradiated by SER-TAIN® process, sterile-filtered, suitable for cell culture
12303C
Fetal Bovine Serum, USDA approved countries, sterile-filtered, cell culture tested
F1051
Fetal Bovine Serum, Canada origin, sterile-filtered, suitable for cell culture
12003C
Fetal Bovine Serum, Australia origin, USDA approved, sterile-filtered, suitable for cell culture
12007C
Fetal Bovine Serum, Australia origin, γ-irradiated, sterile-filtered, suitable for cell culture
12203C
Fetal Bovine Serum, New Zealand origin, sterile-filtered, suitable for cell culture
F0926
Fetal Bovine Serum, USDA approved, sterile-filtered, suitable for cell culture
12306C
Fetal Bovine Serum, non-USA origin, from USDA approved countries, Heat Inactivated, sterile-filtered, suitable for cell culture
12006C
Fetal Bovine Serum, Australia origin, USDA approved, Heat Inactivated, suitable for cell culture
12207C
Fetal Bovine Serum, New Zealand origin, cell culture tested
F9423
Fetal Bovine Serum, Australia origin, suitable for, USDA approved, sterile-filtered, suitable for cell culture, suitable for hybridoma
F3297
Fetal Bovine Serum, Australia origin, γ-irradiated, cell culture tested
12105C
Fetal Bovine Serum, Dialyzed by ultrafiltration against 0.15M NaCl, USA origin, sterile-filtered, cell culture tested
12103C
Fetal Bovine Serum, USA origin, cell culture tested
12106C
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture
F6765
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture
F7305
Fetal Bovine Serum, Mouse Embryonic Stem Cell Screened, suitable for cell culture, sterile-filtered, USDA Approved
121398
Giemsa stain, certified by the Biological Stain Commission
C14H14ClN3S
G4507
Giemsa stain, technical grade, used as a blood stain
C14H14ClN3S
G9641
Giemsa stain, powder, BioReagent, suitable for cell culture
C14H14ClN3S
G5637
Giemsa stain, certified by the Biological Stain Commission
C14H14ClN3S
11700
Giemsa stain, for microscopy (Bact., Hist.)
C14H14ClN3S