EMAIL THIS PAGE TO A FRIEND
Alternative medicine review : a journal of clinical therapeutic
Potential physiological importance of pyrroloquinoline quinone.
PMID 19803551
Abstract
Pyrroloquinoline quinone (PQQ) is a novel biofactor for which a proposition can be made for physiological importance. PQQ was first recognized as an enzyme cofactor in bacteria. It has recently been tentatively identified as a component of interstellar dust. Thus, PQQ may have been present throughout early biological conception and evolution. PQQ is also a potent plant growth factor. Consequently, for animals and humans, there has been constant exposure to PQQ. In animals, PQQ is reported to participate in a range of biological functions with apparent survival benefits (e.g., improved neonatal growth and reproductive performance). There are also benefits from PQQ supplementation related to cognitive, immune, and antioxidant functions, as well as protection from cardiac and neurological ischemic events. Although PQQ is not currently viewed as a vitamin, its involvement in cell signaling pathways, particularly those important to mitochondriogenesis in experimental animal models, may eventually provide a rationale for defining PQQ as vital to life. For humans, such evidence suggests there may be similar parallels or benefits from improving PQQ status.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7783
Pyrroloquinoline quinone, ≥95.0% (HPLC)
C14H6N2O8
64682
Pyrroloquinoline quinone, ≥97.0% (HPLC)
C14H6N2O8