EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
7-Hydroxy-benzopyran-4-one derivatives: a novel pharmacophore of peroxisome proliferator-activated receptor alpha and -gamma (PPARalpha and gamma) dual agonists.
PMID 19807106
Abstract
Design, synthesis, and in vitro bioevaluation of a new class of potential dual PPARalpha and gamma agonists discovered through a structure-driven design paradigm are described. The 7-hydroxy-benzopyran-4-one moiety (includes flavones, flavanones, and isoflavones) is the key pharmacophore of these novel molecules, exhibiting similarity to the core structure of both fibrates and thiazolidinediones. New lead PPAR ligands were identified from "natraceuticals" and synthetic analogues. In total, 77 molecules, including chalcones, flavones, flavanones, isoflavones, and pyrazole derivatives, were screened and structure-activity relationship studies of the dual agonists undertaken. Compounds 68, 70, 72, and 76 were identified as novel and potent dual PPARalpha and gamma agonists. These novel molecules may have the potential to be the future leads in PPAR-related disorders, including type II diabetes mellitus and metabolic syndrome.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C80105
Chrysin, 97%
C15H10O4
D7321
Diosmetin
C16H12O6