EMAIL THIS PAGE TO A FRIEND
Advances in biochemical psychopharmacology
Zolpidem and alpidem: two imidazopyridines with selectivity for omega 1- and omega 3-receptor subtypes.
PMID 1981304
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1862
Alpidem, ≥98% (HPLC), powder
C21H23Cl2N3O