EMAIL THIS PAGE TO A FRIEND
Journal of cell science
Identification of targets of Prox1 during in vitro vascular differentiation from embryonic stem cells: functional roles of HoxD8 in lymphangiogenesis.
PMID 19825936
Abstract
During lymphatic development, Prox1 plays central roles in the differentiation of blood vascular endothelial cells (BECs) into lymphatic endothelial cells (LECs), and subsequently in the maturation and maintenance of lymphatic vessels. However, the molecular mechanisms by which Prox1 elicits these functions remain to be elucidated. Here, we identified FoxC2 and angiopoietin-2 (Ang2), which play important roles in the maturation of lymphatic vessels, as novel targets of Prox1 in mouse embryonic-stem-cell-derived endothelial cells (MESECs). Furthermore, we found that expression of HoxD8 was significantly induced by Prox1 in MESECs, a finding confirmed in human umbilical vein endothelial cells (HUVECs) and human dermal LECs (HDLECs). In mouse embryos, HoxD8 expression was significantly higher in LECs than in BECs. In a model of inflammatory lymphangiogenesis, diameters of lymphatic vessels of the diaphragm were increased by adenovirally transduced HoxD8. We also found that HoxD8 induces Ang2 expression in HDLECs and HUVECs. Moreover, we found that HoxD8 induces Prox1 expression in HUVECs and that knockdown of HoxD8 reduces this expression in HDLECs, suggesting that Prox1 expression in LECs is maintained by HoxD8. These findings indicate that transcriptional networks of Prox1 and HoxD8 play important roles in the maturation and maintenance of lymphatic vessels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN10266 CompoZr® Knockout ZFN Kit, Human HOXD8 (NM_019558)