EMAIL THIS PAGE TO A FRIEND
Planta medica
Anti-leishmanial lindenane sesquiterpenes from Hedyosmum angustifolium.
PMID 19830657
Abstract
The aim of this work is the isolation of anti-leishmanial compounds from the ethyl acetate extracts of the bark of HEDYOSMUM ANGUSTIFOLIUM. We have successfully isolated and characterized five sesquiterpenes: one new compound (oxyonoseriolide, 1), one compound isolated for the first time from a natural source (hedyosmone, 2), and three known sesquiterpenes (onoseriolide, 3; chloranthalactone A, 4; and spathulenol, 5) that had not been previously isolated from H. ANGUSTIFOLIUM. The biological activities of 1- 5 showed that onoseriolide ( 3) was the most active compound against axenic amastigotes from LEISHMANIA AMAZONENSIS and L. INFANTUM. Moreover, it was still active on the intramacrophagic amastigotes of L. INFANTUM. The isolated compounds have also been tested on PLASMODIUM FALCIPARUM and against various mammalian cell lines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6628
Chloroquine diphosphate salt, solid, ≥98%
C18H26ClN3 · 2H3PO4
P0547
Pentamidine isethionate salt, powder
C23H36N4O10S2