EMAIL THIS PAGE TO A FRIEND
BMC genomics
Identification of novel androgen-responsive genes by sequencing of LongSAGE libraries.
PMID 19832994
Abstract
The development and maintenance of the prostate is dependent on androgens and the androgen receptor. The androgen pathway continues to be important in prostate cancer. Here, we evaluated the transcriptome of prostate cancer cells in response to androgen using long serial analysis of gene expression (LongSAGE) libraries. There were 131 tags (87 genes) that displayed statistically significant (p