EMAIL THIS PAGE TO A FRIEND
Chemical Society reviews
Domino reactions of rhodium(II) carbenoids for alkaloid synthesis.
PMID 19847342
Abstract
In this tutorial review, the rhodium(II)-catalyzed domino reactions of alpha-diazo carbonyl compounds are summarized. The article will emphasize some of the more recent synthetic applications of the rhodium carbenoid cyclization/cycloaddition domino cascade for alkaloid synthesis. The many structurally diverse and highly successful examples of both oxa and azapolycyclic ring formation cited in this tutorial review clearly demonstrate that the domino cyclization/cycloaddition cascade of metallo carbenoids has evolved as an important strategy for the synthesis of nitrogen containing natural products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
357340
Rhodium, foil, thickness 0.025 mm, 99.9% trace metals basis
Rh
204218
Rhodium, powder, 99.95% trace metals basis
Rh
267368
Rhodium, sponge, 99.9% trace metals basis
Rh
277916
Rhodium, wire, diam. 1.0 mm, 99.9% trace metals basis
Rh
GF94930667
Rhodium, microfoil, disks, 10mm, thinness 0.25μm, specific density 310μg/cm2, permanent mylar 3.5μm support, 99.99%
Rh
GF64213680
Rhodium, wire reel, 0.05m, diameter 0.12mm, as drawn, 99.9%
Rh
GF37380089
Rhodium, wire reel, 0.2m, diameter 0.25mm, hard, 99.9%
Rh
GF45819995
Rhodium, wire reel, 0.05m, diameter 0.25mm, hard, 99.9%
Rh
GF83497652
Rhodium, wire reel, 0.05m, diameter 1.0mm, as drawn, 99.9%
Rh
GF26710680
Rhodium, wire reel, 0.1m, diameter 0.12mm, as drawn, 99.9%
Rh
GF27499045
Rhodium, wire reel, 0.1m, diameter 0.25mm, hard, 99.9%
Rh
GF06539979
Rhodium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9%
Rh
GF37207166
Rhodium, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.9%
Rh
GF38835737
Rhodium, wire reel, 0.2m, diameter 0.12mm, as drawn, 99.9%
Rh
GF17496973
Rhodium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.9%
Rh
GF54466937
Rhodium, wire reel, 0.5m, diameter 0.12mm, as drawn, 99.9%
Rh
GF72510364
Rhodium, wire reel, 0.5m, diameter 0.25mm, hard, 99.9%
Rh
GF44569907
Rhodium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.9%
Rh
GF80841146
Rhodium, foil, 25x25mm, thickness 0.15mm, as rolled, 99.9%
Rh
GF40539405
Rhodium, foil, 25x25mm, thickness 0.1mm, as rolled, 99.9%
Rh
GF20569697
Rhodium, foil, 12mm disks, thickness 0.5mm, as rolled, 99.9%
Rh
GF61934429
Rhodium, foil, 12mm disks, thickness 1.0mm, as rolled, 99.9%
Rh
GF36379345
Rhodium, foil, light tested, 15x15mm, thickness 0.05mm, as rolled, 99.9%
Rh
GF37647142
Rhodium, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, x-ray quality, 99.9%
Rh
GF57675561
Rhodium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.9%
Rh
GF12568013
Rhodium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, x-ray quality, 99.9%
Rh
GF96356559
Rhodium, foil, light tested, 25x25mm, thickness 0.075mm, as rolled, 99.9%
Rh
GF04978963
Rhodium, foil, not light tested, 10x10mm, thickness 0.012mm, as rolled, 99.9%
Rh
GF17514312
Rhodium, foil, not light tested, 25x25mm, thickness 0.00025mm, permanent polyester support, 99.9%
Rh
GF35442727
Rhodium, foil, not light tested, 25x25mm, thickness 0.0005mm, permanent polyester support, 99.85%
Rh