EMAIL THIS PAGE TO A FRIEND
Pharmazie in unserer Zeit
[Agents for the treatment of leishmaniasis. Antimony, and more].
PMID 19862714
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
452343
Antimony, beads, 1-5 mm, low oxide, 99.999% trace metals basis
Sb
266604
Antimony, beads, 1-2 mm, 99.999% trace metals basis
Sb
266329
Antimony, powder, -100 mesh, 99.5% trace metals basis
Sb
GF72464834
Antimony, foil, not light tested, 100x100mm, thickness 0.015mm, permanent polyester support, 95+%
Sb
GF17616214
Antimony, rod, 6mm, diameter 6mm, 99.999%
Sb
GF72466803
Antimony, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 95+%
Sb
GF20324865
Antimony, rod, 25mm, diameter 2.0mm, 99.99%
Sb
GF76817616
Antimony, foil, 50x50mm, thickness 2mm, hot-pressed
Sb
GF15127961
Antimony, foil, light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 95+%
Sb
GF34540185
Antimony, foil, light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 95+%
Sb
GF97030403
Antimony, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 95+%
Sb
GF90469884
Antimony, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 95+%
Sb
GF45762789
Antimony, foil, not light tested, 100x100mm, thickness 0.05mm, permanent polyester support, 95+%
Sb
GF89164873
Antimony, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 95+%
Sb
GF16935396
Antimony, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 95+%
Sb
GF83770094
Antimony, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 95+%
Sb
GF83929674
Antimony, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 95+%
Sb
GF04779894
Antimony, foil, not light tested, 50x50mm, thickness 0.002mm, permanent polyester support, 95+%
Sb
GF07563203
Antimony, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 95+%
Sb
GF66083362
Antimony, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 95+%
Sb
GF46574220
Antimony, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 95+%
Sb
GF16928310
Antimony, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 95+%
Sb
GF87764013
Antimony, microfoil, 25x25mm, thinness 0.5μm, specific density 334.5μg/cm2, 6 micron aluminum permanent support, 95+%
Sb
GF70501788
Antimony, microfoil, 50x50mm, thinness 0.5μm, specific density 334.5μg/cm2, 6 micron aluminum permanent support, 95+%
Sb
GF59022592
Antimony, rod, 15mm, diameter 2.0mm, 99.99%
Sb
GF96258437
Antimony, rod, 20mm, diameter 4.7mm, 99.999%
Sb
GF87920343
Antimony, rod, 50mm, diameter 2.0mm, 99.99%
Sb
GF01771118
Antimony, rod, 50mm, diameter 4.7mm, 99.999%
Sb
GF97807427
Antimony, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 95+%
Sb