EMAIL THIS PAGE TO A FRIEND
Journal of natural products
The lecanindoles, nonsteroidal progestins from the terrestrial fungus Verticillium lecanii 6144.
PMID 19863083
Abstract
Four new indolosesquiterpenes, lecanindoles A-D (1-4), were isolated from fermentations of the terrestrial fungus Verticillium lecanii 6144. The structures of compounds 1-4 were elucidated from analysis of spectroscopic data. Compound 2 was reduced to give 4 and its isomer 5. Compound 4 was found to be a potent and selective progesterone receptor agonist with an EC50 of 1.1 +/- 0.4 nM in a cell-based luciferase reporter assay.
Related Materials