EMAIL THIS PAGE TO A FRIEND
Journal of drug targeting
Gelatin-based delivery systems for cancer gene therapy.
PMID 19863194
Abstract
Gelatin is a natural, biocompatible, nontoxic, edible, and inexpensive macromolecule. These properties result in its wide application in pharmaceutical, medical, and cosmetic products. Recently, it has been used for the delivery of such gene therapeutic entities as plasmid DNA. This review discusses the in vivo and in vitro studies using gelatin for delivery of therapeutic genes to cancerous cells. Recent studies show that present cancer gene therapy using gelatin is lacking in both efficiency and specificity in comparison with viral vectors, whereas complexes of therapeutic DNA with modified gelatin possibly offer a safe and efficient strategy for systemic administration of therapeutic genes to solid tumors compared to injection of naked plasmid DNA. The future of these promising approaches lies in the development of better techniques for preparing gelatin-gene complexes with the aim of a gelatin-based cancer gene therapy with comparable efficiency to viral vectors but with the added advantage of biosafety for patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G9539
FreAlagin M gelatin, lyophilized powder
G9382
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
G9391 Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
G6650
Gelatin from bovine skin, Type B
G7765 Gelatin from cold water fish skin, 40-50% in H2O
G7041 Gelatin from cold water fish skin, BioReagent, solid
G2500 Gelatin from porcine skin, gel strength 300, Type A
G6144
Gelatin from porcine skin, gel strength 90-110 g Bloom, Type A
G1890 Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, for electrophoresis, suitable for cell culture
G8150
Gelatin from porcine skin, for electrophoresis, Type A
G2625
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
G9136 Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
04055 Gelatin from porcine skin, powder, for microbiology
48720 Gelatin from porcine skin, for microbiology, low gel strength
48722 Gelatin from porcine skin, medium gel strength, for microbiology
48724 Gelatin from porcine skin, high gel strength, for microbiology, for microbiology
39465 Gelatin from porcine skin, for microbiology, ultrahigh gel strength
G1393 Gelatin solution, Type B, 2% in H2O, tissue culture grade, sterile, BioReagent, suitable for cell culture
G0411 Prionex® Highly purified Type A, aqueous solution