EMAIL THIS PAGE TO A FRIEND
Experimental physiology
Role of matrix metalloproteinase-2 in the cardioprotective effect of ischaemic postconditioning.
PMID 19880538
Abstract
The activation of matrix metalloproteinases (MMPs) contributes to myocardial injury at the onset of reperfusion; however, their role in ischaemic postconditioning is unknown. The aim of the present study was to examine the effects of ischaemic postconditioning on MMP activity in isolated rabbit hearts. The isolated rabbit hearts were subjected to 30 min of global ischaemia followed by 180 min of reperfusion (I/R group; n = 8). In the ischaemic postconditioning group (n = 8), a postconditioning protocol was performed (2 cycles of 30 s reperfusion-ischaemia). In other experiments, we added doxycycline, an MMP inhibitor, at 25 (n = 7) or 50 micromol l(1) (n = 8) during the first 2 min of reperfusion. Coronary effluent and left ventricular tissue were collected during pre-ischaemic conditions and at different times during the reperfusion period to measure MMP-2 activity and cardiac protein nitration. We evaluated ventricular function and infarct size. In the I/R group, infarct size was 32.1 +/- 5.2%; Postcon reduced infarct size to 9.5 +/- 3.8% (P < 0.05) and inhibited MMP-2 activity during reperfusion. The administration of doxycycline at 50 micromol l(1) inhibited MMP-2 activity and cardiac protein nitration and reduced the infarct size to 9.7 +/- 2.8% (P < 0.05). A lower dose of doxycycline (25 micromol l(1)) failed to inhibit MMP-2 activity and did not modify the infarct size. Our results strongly suggest that ischaemic postconditioning may exert part of its cardioprotective effects through the inhibition of MMP-2 activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G8150
Gelatin from porcine skin, for electrophoresis, Type A