EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Biologic activities of the TolC-like protein of Neisseria meningitidis as assessed by functional complementation in Escherichia coli.
PMID 19884363
Abstract
Neisseria meningitidis can produce a TolC-like protein needed for secretion of FrpC but not efflux of antimicrobials. We now report that expression of the meningococcal tolC gene in a TolC-deficient strain of Escherichia coli can restore properties of alpha-hemolysis and antimicrobial resistance known to involve efflux pumps.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48770
Gentian violet, for microscopy (Bact., Hist.)