EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and biological evaluation of dehydroabietic acid derivatives.
PMID 19892441
Abstract
A series of C18-oxygenated derivatives of dehydroabietic acid were synthesized from commercial abietic acid and evaluated for their cytotoxic, antimycotic, and antiviral activities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00010
Abietic acid, technical, ~75% (GC)
C20H30O2
SMB00089
Dehydroabietic acid, ≥95% (LC/MS-ELSD)
C20H28O2