EMAIL THIS PAGE TO A FRIEND
Tennessee medicine : journal of the Tennessee Medical Association
Tumor lysis syndrome following chlorambucil therapy for chronic lymphocytic leukemia: case report and review of the literature.
PMID 19899317