EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and in vivo evaluation of 3,4-disubstituted gababutins.
PMID 19910190
Abstract
A range of 3,4-alkylated five-membered ring derivatives of gabapentin were synthesised. One compound (21) had an excellent level of potency against alpha(2)delta and was profiled in in vivo models of pain and anxiety.