EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and inhibitory activity against human monoamine oxidase of N1-thiocarbamoyl-3,5-di(hetero)aryl-4,5-dihydro-(1H)-pyrazole derivatives.
PMID 19926363
Abstract
A series of N1-thiocarbamoyl-3,5-di(hetero)aryl-4,5-dihydro-(1H)-pyrazole derivatives has been synthesized and assayed for their ability to inhibit the activity of the A and B isoforms of human monoamine oxidase (hMAO). Some of these compounds were endowed with a selective inhibitory activity against hMAO-B in the micromolar range. The most active of the series is the compound 13, N1-thiocarbamoyl-3-(fur-2'-yl)-5-(4'-fluoro-phenyl)-4,5-dihydro-(1H)-pyrazole, with IC(50) 2.75+/-0.81muM value and selectivity ratio of 25, which is the best candidate for further investigations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
58240
Isatin, for spectrophotometric det. of proline and thiophene, ≥99.0%
C8H5NO2
114618
Isatin
C8H5NO2