EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Activity and toxicity of farnesol towards Candida albicans are dependent on growth conditions.
PMID 19933803
Abstract
Farnesol interacts with Candida albicans as both a quorum-sensing molecule and toxic agent, but confusion abounds regarding which conditions promote these distinct responses. Farnesol sensitivity was measured when inoculum cell history and size, temperature, and growth media were altered. Parameters for farnesol tolerance/sensitivity were defined, validating previous studies and identifying new variables, such as energy availability. This study clearly defines what farnesol concentrations are lethal to C. albicans, based on environmental conditions.
Related Materials