EMAIL THIS PAGE TO A FRIEND
The Urologic clinics of North America
Holmium laser applications of the prostate.
PMID 19942047
Abstract
The high-powered holmium laser is an excellent tool for the surgical treatment of benign prostatic hyperplasia. This article discusses the background of holmium use in the prostate and describes the surgical techniques of holmium laser ablation of the prostate and holmium laser enucleation of the prostate. Operative challenges are reviewed with suggestions as to how to avoid these problems or deal with them when they arise. Surgical outcomes and a thorough literature review are both presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF92551259
Holmium, rod, 50mm, diameter 3.8mm, 99%
Ho
GF12230154
Holmium, rod, 50mm, diameter 6.35mm, cast, 99%
Ho
GF39809863
Holmium, wire reel, 100mm, diameter 0.5mm, as drawn, 99.9%
Ho
GF22997188
Holmium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Ho
GF59427513
Holmium, wire reel, 200mm, diameter 0.5mm, as drawn, 99.9%
Ho
GF74648223
Holmium, wire reel, 200mm, diameter 1.0mm, hard, 99.9%
Ho
GF88732693
Holmium, wire reel, 50mm, diameter 0.5mm, as drawn, 99.9%
Ho
GF13376785
Holmium, rod, 200mm, diameter 3.8mm, 99%
Ho
GF28769541
Holmium, rod, 100mm, diameter 6.35mm, cast, 99%
Ho
GF68907011
Holmium, rod, 100mm, diameter 3.8mm, 99%
Ho
GF31028543
Holmium, powder, 5g, max. particle size 500 micron, 99.9%
Ho
GF78567654
Holmium, powder, 10g, max. particle size 500 micron, 99.9%
Ho
GF29991673
Holmium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Ho
GF26625067
Holmium, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99%
Ho
GF42613006
Holmium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99%
Ho
GF60938635
Holmium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Ho
GF34764666
Holmium, foil, not light tested, 50x50mm, thickness 0.0125mm, as rolled, 99%
Ho
GF78609640
Holmium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Ho
GF49265253
Holmium, foil, not light tested, 25x25mm, thickness 0.0125mm, as rolled, 99%
Ho
GF07383977
Holmium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Ho
GF85502640
Holmium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Ho
GF89418224
Holmium, foil, 25x25mm, thickness 0.1mm, as rolled, 99%
Ho
GF19613684
Holmium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Ho
GF38519376
Holmium, foil, not light tested, 100x100mm, thickness 0.0125mm, as rolled, 99%
Ho