EMAIL THIS PAGE TO A FRIEND
Epigenetics & chromatin
Core promoter acetylation is not required for high transcription from the phosphoenolpyruvate carboxylase promoter in maize.
PMID 19954517
Abstract
Acetylation of promoter nucleosomes is tightly correlated and mechanistically linked to gene activity. However, transcription is not necessary for promoter acetylation. It seems, therefore, that external and endogenous stimuli control histone acetylation and by this contribute to gene regulation. Photosynthetic genes in plants are excellent models with which to study the connection between stimuli and chromatin modifications because these genes are strongly expressed and regulated by multiple stimuli that are easily manipulated. We have previously shown that acetylation of specific histone lysine residues on the photosynthetic phosphoenolpyruvate carboxylase (Pepc) promoter in maize is controlled by light and is independent of other stimuli or gene activity. Acetylation of upstream promoter regions responds to a set of other stimuli which include the nutrient availability of the plant. Here, we have extended these studies by analysing histone acetylation during the diurnal and circadian rhythm of the plant. We show that histone acetylation of individual lysine residues is removed from the core promoter before the end of the illumination period which is an indication that light is not the only factor influencing core promoter acetylation. Deacetylation is accompanied by a decrease in gene activity. Pharmacological inhibition of histone deacetylation is not sufficient to prevent transcriptional repression, indicating that deacetylation is not controlling diurnal gene regulation. Variation of the Pepc promoter activity during the day is controlled by the circadian oscillator as it is maintained under constant illumination for at least 3 days. During this period, light-induced changes in histone acetylation are completely removed from the core promoter, although the light stimulus is continuously applied. However, acetylation of most sites on upstream promoter elements follows the circadian rhythm. Our results suggest a central role of upstream promoter acetylation in the quantitative regulation of gene expression in this model gene. Induced core promoter acetylation is dispensable for the highest gene expression in the diurnal and circadian rhythm.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-353
Anti-acetyl-Histone H3 (Lys14) Antibody, Anti-acetyl-Histone H3 (Lys14) Antibody is a rabbit polyclonal antibody for detection of acetyl-Histone H3 (Lys14) also known as H3K14Ac, Histone H3 (acetyl K14) & has been validated in ChIP, WB, ChIP-seq, DB, Mplex.
07-354
Anti-acetyl-Histone H3 (Lys18) Antibody, Use Anti-acetyl-Histone H3 (Lys18) Antibody (Rabbit Polyclonal Antibody) validated in ChIP, WB to detect acetyl-Histone H3 (Lys18) also known as H3K18Ac, Histone H3 (acetyl K18).
07-355
Anti-acetyl-Histone H3 (Lys23) Antibody, Use Anti-acetyl-Histone H3 (Lys23) Antibody (Rabbit Polyclonal Antibody) validated in ChIP, WB to detect acetyl-Histone H3 (Lys23) also known as H3K23Ac, Histone H3 (acetyl K23).
07-360
Anti-acetyl-Histone H3 (Lys27) Antibody, Anti-acetyl-Histone H3 (Lys27) Antibody is a rabbit polyclonal antibody for detection of Histone H3 acetylated on lysine 27. Also known as Anti-H3K27ac this antibody is published in peer reviewed journals and is specificity verified by dot blot (DB) and validated in ChIP, ChIP-seq, WB, DB, Mplex.
07-352
Anti-acetyl-Histone H3 (Lys9) Antibody, Anti-acetyl-Histone H3 (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of acetyl-Histone H3 (Lys9) also known as H3K9Ac, Histone H3 (acetyl K9) and has been published and validated in ChIP, WB, Mplex.
07-329
Anti-acetyl-Histone H4 (Lys16) Antibody, Anti-acetyl-Histone H4 (Lys16) Antibody is a rabbit polyclonal antibody for detection of acetyl-Histone H4 (Lys16) also known as H4K16Ac, Histone H4 (acetyl K16 & has been published and validated in ChIP, WB, Mplex, PIA, DB, ChIP-seq.
07-327
Anti-acetyl-Histone H4 (Lys5) Antibody, Use Anti-acetyl-Histone H4 (Lys5) Antibody (Rabbit Polyclonal Antibody) validated in WB, DB, ICC, ChIP-seq, ChIP to detect acetyl-Histone H4 (Lys5) also known as H4K5Ac, Histone H4 (acetyl K5).
Z0876
trans-Zeatin, BioReagent, plant cell culture tested, ≥97%
C10H13N5O