EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Two novel neo-clerodane diterpenoids from Scutellaria barbata.
PMID 19962306
Abstract
Two novel neo-clerodane diterpenoids, barbatellarines A (1) and B (2), were isolated from the whole plants of Scutellaria barbata, along with the known compound scutebarbatine F (3). The chemical structures and relative stereochemistry of the isolated compounds were established by NMR (1D and 2D) and mass spectroscopic analyses. Compounds 2 and 3 were evaluated for in vitro cytotoxic activity against the HL-60 (human leukemia), MCF7 (human breast cancer), and LLC (Lewis lung carcinoma) cancer cell lines. Compound 2 exhibited weak cytotoxic activity against HL-60 cells, with an IC(50) value of 41.4microM.