EMAIL THIS PAGE TO A FRIEND
Brain research reviews
Pharmacological and genetic approaches to study connexin-mediated channels in glial cells of the central nervous system.
PMID 19963007
Abstract
This review gives an overview of connexin expression in glial cells of the central nervous system, the different modes of connexin action, including gap junctional channels and hemichannels, as well as the available methodologies to measure their activity. We summarize the strengths and limitations of current pharmacological and genetic approaches to interfere with connexin channel functions. We outline new avenues not only to study specific mechanisms by which connexins exert these functions but also to selectively investigate well-defined coupling compartments among glial networks.
Related Materials