EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Evidence for an involvement of the brain cholecystokinin B receptor in anxiety.
PMID 1996314
Abstract
The effect of neuropeptide cholecystokinin (CCK) receptor agonists and antagonists was examined in the rat elevated X-maze model of anxiety. The selective CCK-B receptor antagonists CI-988 (PD 134308) and L-365,260 produced anxiolytic-like effects, whereas MK-329, a CCK-A receptor antagonist, was respectively less potent by factors of 313 and 200. The intracerebroventricular administration of the nonselective CCK receptor agonist caerulein or the selective CCK-B receptor agonist pentagastrin increased dose dependently the level of anxiety. CI-988 dose dependently antagonized the anxiogenic response to pentagastrin but not that induced by pentylenetetrazol. These results strongly suggest that activation of the brain CCK-B receptor induces anxiety and that selective antagonists of this receptor represent a separate class of anxiolytic agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1636
Pentagastrin, ≥95% (HPLC), powder
C37H49N7O9S