EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
A toxic disinfection by-product, 2,6-dichloro-1,4-benzoquinone, identified in drinking water.
PMID 20025006
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
431982
2,6-Dichloro-1,4-benzoquinone, 98%
C6H2Cl2O2