EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Combined ramR mutation and presence of a Tn1721-associated tet(A) variant in a clinical isolate of Salmonella enterica serovar Hadar resistant to tigecycline.
PMID 20028818
Abstract
A Salmonella enterica serovar Hadar strain resistant to tigecycline (MIC, 16 microg/ml) was isolated. Molecular characterization revealed the presence of a plasmid-borne tet(A) variant associated with Tn1721 mediating a rise of the MIC for tigecycline when transferred to Escherichia coli. Additionally, a truncating mutation in ramR was detected. Transformation with wild-type ramR but not with the mutated ramR lowered the MIC for tigecycline. Characterization of this Salmonella isolate implicates ramR in resistance to tigecycline.