EMAIL THIS PAGE TO A FRIEND
Circulation. Cardiovascular genetics
Determination of paraoxonase 1 status without the use of toxic organophosphate substrates.
PMID 20031556
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
432881
4-(Chloromethyl)phenyl acetate, 98%
C9H9ClO2