EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
1-Arylsulfonyl-2-(pyridylmethylsulfinyl) benzimidazoles as new proton pump inhibitor prodrugs.
PMID 20032890
Abstract
New arylsulfonyl proton pump inhibitor (PPI) prodrug forms were synthesized. These prodrugs provided longer residence time of an effective PPI plasma concentration, resulting in better gastric acid inhibition.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
446386
2-(p-Tolylsulfonyl)ethanol, 97%
C9H12O3S