EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
Prenatal stress alters presynaptic marker proteins in the hippocampus of rat pups.
PMID 20035832
Abstract
Exposure to stress during critical periods of an organism's maturation can result in permanent behavioral changes and induced hyper-responsive to aversive stimuli as adult. Hippocampus is a plastic and vulnerable brain structure that is susceptible to damage during aging and repeated stress. The present study examines the effect of maternal restraint stress on the level of GAP-43, pGAP-43 and synaptophysin in the hippocampus of rat pups. Prenatal stress (PS) causes a significant increase of GAP-43 and pGAP-43 (p
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2505
Corticosterone, ≥92%
C21H30O4