EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Acquisition of flucytosine, azole, and caspofungin resistance in Candida glabrata bloodstream isolates serially obtained from a hematopoietic stem cell transplant recipient.
PMID 20038613
Abstract
We describe the acquisition of flucytosine, azole, and caspofungin resistance in sequential Candida glabrata bloodstream isolates collected from a bone marrow transplant patient with clinical failure. Point mutations in C. glabrata FUR1 (CgFUR1) and CgFKS2 and overexpression of CgCDR1 and CgCDR2 were observed in resistant isolates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F7129
5-Fluorocytosine, nucleoside analog
C4H4FN3O