EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Getting started with yeast.
PMID 2005794
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
84605 SD Agar, for microbiology