EMAIL THIS PAGE TO A FRIEND
Mutation research
The genetic toxicology of styrene and styrene oxide.
PMID 2005936
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S5006
Styrene oxide, 97%
C8H8O
W513504
Styrene oxide, ≥97%
C8H8O