EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Comparative in vitro activities of nemonoxacin (TG-873870), a novel nonfluorinated quinolone, and other quinolones against clinical isolates.
PMID 20065058
Abstract
The in vitro antibacterial activities of nemonoxacin (TG-873870), a novel nonfluorinated quinolone, against 770 clinical isolates were investigated. Nemonoxacin (tested as its malate salt, TG-875649) showed better in vitro activity than ciprofloxacin and levofloxacin against different species of staphylococci, streptococci, and enterococci, Neisseria gonorrhoeae, and Haemophilus influenzae. The in vitro activity of TG-875649 was also comparable to or better than that of moxifloxacin against these pathogens, which included ciprofloxacin-resistant, methicillin-resistant Staphylococcus aureus and levofloxacin-resistant Streptococcus pneumoniae.