EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Isosorbide-2-benzyl carbamate-5-salicylate, a peripheral anionic site binding subnanomolar selective butyrylcholinesterase inhibitor.
PMID 20067290
Abstract
Isosorbide-2-benzyl carbamate-5-benzoate is a highly potent and selective BuChE inhibitor. Meanwhile, isosorbide-2-aspirinate-5-salicylate is a highly effective aspirin prodrug that relies on the salicylate portion to interact productively with human BuChE. By integrating the salicylate group into the carbamate design, we have produced isosorbide-2-benzyl carbamate-5-salicylate, an inhibitor of high potency (150 pM) and selectivity for human BuChE over AChE (666000) and CES2 (23000). Modeling and mutant studies indicate that it achieves its exceptional potency because of an interaction with the polar D70/Y332 cluster in the PAS of BuChE in addition to pseudosubstrate interactions with the active site.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E8375
Eserine
C15H21N3O2