EMAIL THIS PAGE TO A FRIEND
Molecular & cellular proteomics : MCP
Integrated protein array screening and high throughput validation of 70 novel neural calmodulin-binding proteins.
PMID 20068228
Abstract
Calmodulin is an essential regulator of intracellular processes in response to extracellular stimuli mediated by a rise in Ca(2+) ion concentration. To profile protein-protein interactions of calmodulin in human brain, we probed a high content human protein array with fluorophore-labeled calmodulin in the presence of Ca(2+). This protein array contains 37,200 redundant proteins, incorporating over 10,000 unique human neural proteins from a human brain cDNA library. We designed a screen to find high affinity (K(D) < or = 1 microm) binding partners of calmodulin and identified 76 human proteins from all intracellular compartments of which 72 are novel. We measured the binding kinetics of 74 targets with calmodulin using a high throughput surface plasmon resonance assay. Most of the novel calmodulin-target complexes identified have low dissociation rates (k(off) < or = 10(-3) s(-1)) and high affinity (K(D)