EMAIL THIS PAGE TO A FRIEND
The European journal of neuroscience
In vivo labelling of hippocampal beta-amyloid in triple-transgenic mice with a fluorescent acetylcholinesterase inhibitor released from nanoparticles.
PMID 20092557
Abstract
The drastic loss of cholinergic projection neurons in the basal forebrain is a hallmark of Alzheimer's disease (AD), and drugs most frequently applied for the treatment of dementia include inhibitors of the acetylcholine-degrading enzyme acetylcholinesterase (AChE). This protein is known to act as a ligand of beta-amyloid (Abeta) in senile plaques, a further neuropathological sign of AD. Recently, we have shown that the fluorescent, heterodimeric AChE inhibitor PE154 allows for the histochemical staining of cortical Abeta plaques in triple-transgenic (TTG) mice with age-dependent beta-amyloidosis and tau hyperphosphorylation, an established animal model for aspects of AD. In the present study, we have primarily demonstrated the targeting of Abeta-immunopositive plaques with PE154 in vivo for 4 h up to 1 week after injection into the hippocampi of 13-20-month-old TTG mice. Numerous plaques, double-stained for PE154 and Abeta-immunoreactivity, were revealed by confocal laser-scanning microscopy. Additionally, PE154 targeted hippocampal Abeta deposits in aged TTG mice after injection of carboxylated polyglycidylmethacrylate nanoparticles delivering the fluorescent marker in vivo. Furthermore, biodegradable core-shell polystyrene/polybutylcyanoacrylate nanoparticles were found to be suitable, alternative vehicles for PE154 as a useful in vivo label of Abeta. Moreover, we were able to demonstrate that PE154 targeted Abeta, but neither phospho-tau nor reactive astrocytes surrounding the plaques. In conclusion, nanoparticles appear as versatile carriers of AChE inhibitors and other promising drugs for the treatment of AD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0624 PE 154, ≥97% (HPLC)
C35H35N5O4