EMAIL THIS PAGE TO A FRIEND
Cardiovascular research
The 5-lipoxygenase/leukotriene pathway in preclinical models of cardiovascular disease.
PMID 20093252
Abstract
Leukotrienes (LTs) derived from 5-lipoxygenase (5-LO) activity are most widely known for their actions during acute inflammation and asthma. 5-LO/LT pathway involvement in cardiovascular disease (CVD) pathogenesis has come to the forefront based on provocative human genetic/population and animal studies leading to the hypothesis that this pathway promotes atherosclerosis, abdominal aortic aneurysm, and myocardial infarction/reperfusion injury via increased leucocyte chemotaxis, vascular inflammation and enhanced permeability, and subsequent tissue/matrix degeneration. A series of pre-clinical studies have tested this hypothesis by means of genetic or pharmacological inhibition of either the LT biosynthesis axis (5-LO, 5-LO-activating protein, LTA(4) hydrolase, LTC(4) synthase) or the cognate LT receptors. Here, we summarize, compare, and analyse these animal studies and relate their findings to human disease pathogenesis. We draw a complex picture of 5-LO/LT participation in cardiovascular disorders, which is further complicated by marked differences between species. Moreover, we discuss how the cytokine footprint of the respective pathological conditions determines the expression level and hence, the contribution of components of the pathway to the overall disease state. Current knowledge implies a role for 5-LO and LTs during the early/acute phase of CVD, but our understanding of a putative 5-LO/LT involvement in more advanced stages of CVD is limited, thereby preventing simple extrapolation of findings from animal studies to humans.