EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
HDAC4 represses matrix metalloproteinase-13 transcription in osteoblastic cells, and parathyroid hormone controls this repression.
PMID 20097749
Abstract
Parathyroid hormone (PTH) is a hormone regulating bone remodeling through its actions on both bone formation and bone resorption. Previously we reported that PTH induces matrix metalloproteinase-13 (MMP-13) transcription in osteoblastic cells. Here, we show that histone deacetylase 4 (HDAC4) interacts with Runx2, binds the MMP-13 promoter, and suppresses MMP-13 gene transcription in the rat osteoblastic cell line, UMR 106-01. PTH induces the rapid cAMP-dependent protein kinase-dependent release of HDAC4 from the MMP-13 promoter and subsequent transcription of MMP-13. Knock-out of HDAC4 either by siRNA in vitro or by gene deletion in vivo leads to an increase in MMP-13 expression, and overexpression of HDAC4 decreases the PTH induction of MMP-13. All of these observations indicate that HDAC4 represses MMP-13 gene transcription in bone. Moreover, PTH stimulates HDAC4 gene expression and enzymatic activity at times corresponding to the reassociation of HDAC4 with the MMP-13 promoter and a decline in its transcription. Thus, HDAC4 is a basal repressor of MMP-13 transcription, and PTH regulates HDAC4 to control MMP-13 promoter activity. These data identify a novel and discrete mechanism of regulating HDAC4 levels and, subsequently, gene expression.