EMAIL THIS PAGE TO A FRIEND
FEBS letters
Intrahepatic cholesterol influences progression, inhibition and reversal of non-alcoholic steatohepatitis in hyperlipidemic mice.
PMID 20114046
Abstract
Hepatic inflammation is the key factor in non-alcoholic steatohepatitis (NASH) and promotes progression to liver damage. We recently identified dietary cholesterol as the cause of hepatic inflammation in hyperlipidemic mice. We now show that hepatic transcriptome responses are strongly dependent on cholesterol metabolism during diet-induced NASH and its inhibition by fenofibrate. Furthermore, we show that, despite doubling hepatic steatosis, pharmacological LXR activation reverses hepatic inflammation, in parallel with reversing hepatic cholesterol levels. Together, the results indicate a prominent role of cholesterol during the development, inhibition and reversal of hepatic inflammation in NASH and reveal potential new therapeutic strategies against NASH.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6020
Fenofibrate, ≥99%, powder
C20H21ClO4