EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Design, synthesis, evaluation and QSAR analysis of N(1)-substituted norcymserine derivatives as selective butyrylcholinesterase inhibitors.
PMID 20137934
Abstract
We synthesized a series of N(1)-substituted norcymserine derivatives 7a-p and evaluated their anti-cholinesterase activities. In vitro evaluation showed that the pyridinylethyl derivatives 7m-o and the piperidinylethyl derivative 7p improved the anti-butyrylcholinesterase activity by approximately threefold compared to N(1)-phenethylnorcymserine (PEC, 2). A quantitative structure-activity relationship (QSAR) study indicated that logS might be a key feature of the improved compounds.