EMAIL THIS PAGE TO A FRIEND
SAAD digest
Propranolol and D-cycloserine as adjunctive medications in reducing dental fear in sedation practice.
PMID 20151608
Abstract
Extensive research and clinical experience have demonstrated the usefulness of sedation in helping fearful patients receive dental treatment, particularly when they have urgent treatment needs. In addition, the efficacy of behavioural programmes for managing dental fears is well established. While often these two approaches are seen as oppositional, our work in Seattle, Morgantown and at King's College London Dental Institute demonstrates the complementarity of the two approaches. Using the example of two compounds, one very familiar, propranolol, and one that has recently become of interest, D-cycloserine, we wish to illustrate the manner in which these medications can be used to enhance behavioural approaches to managing dental anxiety.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6880
D-Cycloserine, powder
C3H6N2O2
C3909
D-Cycloserine, synthetic
C3H6N2O2
30020
D-Cycloserine, ≥96.0% (NT)
C3H6N2O2