EMAIL THIS PAGE TO A FRIEND
Tetrahedron letters
A simple and expedient method for the preparation of N-chlorohydantoins.
PMID 20157342
Abstract
A simple and efficient methodology for the preparation of N-chlorinated hydantoins is presented. These versatile chlorenium sources were isolated in high yield after a simple recrystallization. Among the ten examples are the first chiral N-chlorohydantoins.