EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and evaluation of some new substituted benzothiazepine and benzoxazepine derivatives as anticonvulsant agents.
PMID 20163892
Abstract
A new series of 4-(4'-Hydroxyphenyl)-2-(3-substitutedphenyl)-3-(4-substitutedphenylamino methylene)-2,3-dihydro-1,5-benzothiazepines (3a-3j) and 4-(4'-Hydroxyphenyl)-2-(3-substituted phenyl)-3-(4-substitutedphenylaminomethylene)-2,3-dihydro-1,5-benzoxazepins (3a'-3j') were synthesized and evaluated for their anticonvulsant activity. All these compounds were screened in vivo, for their anticonvulsant activity and acute toxicity. Compound 3g was found to be most potent compound of this series and was compared with the reference drug phenytion sodium. The structures of these compounds have been established by IR, (1)H NMR and (13)C NMR spectroscopic data.