EMAIL THIS PAGE TO A FRIEND
Cancer biology & therapy
Can indole-based extracts prevent colorectal cancer via early apoptotic pathways?
PMID 20168074
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48157 Indole solution, 2000 μg/mL in methanol, certified reference material