EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Discovery of dipeptides with high affinity to the specific binding site for substance P1-7.
PMID 20178322
Abstract
Substance P 1-7 (SP(1-7), H-Arg-Pro-Lys-Pro-Gln-Gln-Phe-OH) is the major bioactive metabolite of substance P. The interest in this heptapeptide originates from the observation that it modulates, and in certain cases opposes the effects of the parent peptide, e.g., the nociceptive effect. The mu-opioid receptor agonist endomorphin-2 (EM-2, H-Tyr-Pro-Phe-Phe-NH(2)) has been found to also interact with the specific binding site of SP(1-7) with only a 10-fold lower affinity compared to the native peptide. Considering the smaller size of EM-2 compared to the target heptapeptide, it was selected as a lead compound in the development of low-molecular-weight ligands to the SP(1-7) binding site. An alanine scan and truncation study led to the unexpected discovery of the dipeptide H-Phe-Phe-NH(2) (K(i) = 1.5 nM), having equal affinity as the endogenous heptapeptide SP(1-7.) Moreover, the studies show that the C-terminal phenylalanine amide is crucial for the affinity of the dipeptide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1883
L-Phenylalaninamide
C9H12N2O