EMAIL THIS PAGE TO A FRIEND
Toxicological sciences : an official journal of the Society of Toxicology
Increased susceptibility of Nrf2-null mice to 1-bromopropane-induced hepatotoxicity.
PMID 20211940
Abstract
1-Bromopropane (1-BP) was introduced as an alternative to ozone-depleting solvents. However, it was found to exhibit neurotoxicity, reproductive toxicity, and hepatotoxicity in rodents and neurotoxicity in human. However, the mechanisms underlying the toxicities of 1-BP remain elusive. The present study investigated the role of oxidative stress in 1-BP-induced hepatotoxicity using nuclear factor erythroid 2-related factor 2 (Nrf2)-null mice. Groups of 24 male Nrf2-null mice and 24 male wild-type (WT) C57BL/6J mice were each divided into three groups of eight and exposed to 1-BP at 0, 100, or 300 ppm for 8 h/day for 28 days by inhalation. Liver histopathology showed significantly larger area of necrosis in Nrf2-null mice relative to WT mice at the same exposure level. Nrf2-null mice also had greater malondialdehyde (MDA) levels, higher ratio of oxidized glutathione/reduced form of glutathione, and lower total glutathione content. The constitutive level and the increase in ratio per exposure level of glutathione S-transferase (GST) activity were lower in the liver of Nrf2-null mice than WT mice. Exposure to 1-BP at 300 ppm increased the messenger RNA levels of heme oxygenase-1 (HO-1), glutamate-cysteine ligase modifier subunit (GcLm), glutamate-cysteine synthetase (GcLc), glutathione reductase, and NAD(P)H: quinone oxidoreductase 1 (NQO1) in WT mice but not in Nrf2-null mice except for GST Yc2. Nrf2-null mice were more susceptible to 1-BP-induced hepatotoxicity. That oxidative stress plays a role in 1-BP hepatotoxicity is deduced from the low expression levels and activities of antioxidant enzymes and high MDA levels in Nrf2-null mice.